Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP50 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093324
Description
NUP50 Polyclonal specifically detects NUP50 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
NUP50 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
50 kDa nucleoporin, MGC39961, NPAP60, NPAP60LNucleoporin Nup50, nuclear pore complex protein Nup50, Nuclear pore-associated protein 60 kDa-like, nuclear pore-associated protein 60L, nucleoporin 50kD, nucleoporin 50kDa | |
Rabbit | |
50 kDa | |
0.1 mL | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Bovine, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Crab-eating macaque, Small-eared galago and Northern whitecheeked gibbon. | |
Store at 4C. Do not freeze. |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunohistochemistry 1:200 - 1:1000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
Q9UKX7 | |
NUP50 | |
The immunogen recognized by this antibody maps to a region between residue 418 and 468 of human nucleoporin 50kDa using the numbering given in entry NP_009103.2 (GeneID 10762). | |
Affinity Purified | |
RUO | |
10762 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction