Learn More
Specifications
Specifications
| Preparation Method | This product is intended for use as a positive control in Western Blot. Each vial of cell lysate contains 100 ug of total protein which should be sufficient for 20-50 reactions. Depending on over-expression level, antibody affinity and detection system, some lysates can go as low as 0.1 μL / load. We recommend starting with 5 ug of cell lysate. Add an equal amount of cell lysate and 2 x SDS Sample buffer and boil the SDS samples for 10 minutes before loading. |
| Quantity | 0.1 mg |
| Host Species | Human |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| Target | NUP50 |
| Synonym | 50 kDa nucleoporin, MGC39961, NPAP60, NPAP60LNucleoporin Nup50, nuclear pore complex protein Nup50, Nuclear pore-associated protein 60 kDa-like, nuclear pore-associated protein 60 L, nucleoporin 50 kD, nucleoporin 50 kDa |
| Gene Symbol | NUP50 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
