Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569833
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569833 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569833 Supplier Invitrogen™ Supplier No. PA569833
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 85%; Rat: 100%.

The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. Pseudogenes of this gene are found on chromosomes 5, 6, and 14. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP50
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NUP50
Gene Accession No. O08587
Gene Alias 1700030K07Rik; 50 kDa nucleoporin; AI413123; CTA-268H5.7; DKFZP564A043; MGC39961; Npap60; NPAP60L; nuclear pore associated protein; nuclear pore associated protein 60 kDa; nuclear pore complex protein Nup50; nuclear pore complex protein Nup50; LOW QUALITY PROTEIN: nuclear pore complex protein Nup50; nuclear pore complex protein nup50-like protein; Nuclear pore-associated protein 60 kDa-like; nuclear pore-associated protein 60L; nucleoporin 50; nucleoporin 50 kDa; Nucleoporin 50 kDa-like protein; nucleoporin 50kD; nucleoporin 50kDa; nucleoporin Nup50; nucleoprotein 50; Nucleoprotein 50kD; Nup50; NUP50 protein; OTTHUMP00000199530; PRO1146; Rtp60; Unknown (protein for MGC:134469)
Gene Symbols NUP50
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of rat NUP50.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25497
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.