Learn More
Invitrogen™ NUP50 Polyclonal Antibody

Description
Recommended positive controls: H1299, Jurkat, Raji, NCI-H929, mouse testis. Predicted reactivity: Mouse (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | NUP50 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.79 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | NUP50 |
| Gene Accession No. | Q9JIH2, Q9UKX7 |
| Gene Alias | 1700030K07Rik; 50 kDa nucleoporin; AI413123; CTA-268H5.7; DKFZP564A043; MGC39961; Npap60; NPAP60L; nuclear pore associated protein; nuclear pore associated protein 60 kDa; nuclear pore complex protein Nup50; nuclear pore complex protein Nup50; LOW QUALITY PROTEIN: nuclear pore complex protein Nup50; nuclear pore complex protein nup50-like protein; Nuclear pore-associated protein 60 kDa-like; nuclear pore-associated protein 60L; nucleoporin 50; nucleoporin 50 kDa; Nucleoporin 50 kDa-like protein; nucleoporin 50kD; nucleoporin 50kDa; nucleoporin Nup50; nucleoprotein 50; Nucleoprotein 50kD; Nup50; NUP50 protein; OTTHUMP00000199530; PRO1146; Rtp60; Unknown (protein for MGC:134469) |
| Gene Symbols | NUP50 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.