Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP54 Monoclonal Antibody (139CT1.1.5)
GREENER_CHOICE

Catalog No. PIMA537776
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537776 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537776 Supplier Invitrogen™ Supplier No. MA537776
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NUP54 Monoclonal Antibody for Western Blot

The nuclear envelope creates distinct nuclear and cytoplasmic compartments in eukaryotic cells. It consists of two concentric membranes perforated by nuclear pores, large protein complexes that form aqueous channels to regulate the flow of macromolecules between the nucleus and the cytoplasm. These complexes are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. This gene encodes a member of the phe-gly repeat-containing nucleoporin subset.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP54
Applications Western Blot
Classification Monoclonal
Clone 139CT1.1.5
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene NUP54
Gene Accession No. Q7Z3B4
Gene Alias 3110079L04Rik; 54 kDa nucleoporin; 54kDa; Nuclear pore complex protein Nup54; nucleoporin 54; nucleoporin 54kD; nucleoporin 54kDa; nucleoporin Nup54; Nucleoporin p54; NUP54
Gene Symbols NUP54
Host Species Mouse
Immunogen NUP54 recombinant protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53371
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.