Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP62 Antibody (M436), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323226
Description
NUP62 Monoclonal antibody specifically detects NUP62 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
NUP62 | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Chicken | |
Purified |
Western Blot, Immunofluorescence | |
M436 | |
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:50 | |
DKFZp547L134,62 kDa nucleoporin, FLJ20822, FLJ43869, IBSNp62, MGC841, nuclear pore glycoprotein p62, nucleoporin 62kD, nucleoporin 62kDa, SNDINucleoporin Nup62 | |
Clone M436 was generated from a recombinant protein containing amino acid residues in the N-terminal region of human nucleoporin p62. This sequence has high homology to similar regions in rat, mouse, and chicken NUP62. [UniProt# P37198] | |
0.1 mL | |
Apoptosis, Cell Biology, Signal Transduction | |
23636 | |
Store at -20°C. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction