Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP62 Antibody (RL31), Biotin, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NB1202736B
Description
NUP62 Monoclonal antibody specifically detects NUP62 in Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
NUP62 | |
Monoclonal | |
Biotin | |
PBS | |
Armenian Hamster | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
Western Blot, Immunofluorescence | |
RL31 | |
Western Blot, Immunofluorescence | |
DKFZp547L134,62 kDa nucleoporin, FLJ20822, FLJ43869, IBSNp62, MGC841, nuclear pore glycoprotein p62, nucleoporin 62kD, nucleoporin 62kDa, SNDINucleoporin Nup62 | |
O-linked glycoprotein-enriched fraction from rat liver nuclear envelopes raised in Armenian hamsters. | |
0.1 mL | |
Apoptosis, Cell Biology, Signal Transduction | |
23636 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction