Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP62 Antibody (RL31), Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NB1202736
Description
NUP62 Monoclonal specifically detects NUP62 in Rat samples. It is validated for Western Blot, Immunofluorescence.Specifications
NUP62 | |
Monoclonal | |
0.5 mg/mL | |
Western Blot 1:200, Immunofluorescence 1:50 | |
DKFZp547L134,62 kDa nucleoporin, FLJ20822, FLJ43869, IBSNp62, MGC841, nuclear pore glycoprotein p62, nucleoporin 62kD, nucleoporin 62kDa, SNDINucleoporin Nup62 | |
Armenian Hamster | |
Protein A purified | |
RUO | |
23636 | |
Rat | |
Purified |
Western Blot, Immunofluorescence | |
RL31 | |
Unconjugated | |
P17955 | |
NUP62 | |
O-linked glycoprotein-enriched fraction from rat liver nuclear envelopes raised in Armenian hamsters. | |
100 μg | |
Primary | |
Detects the nucleoporin p62 O-linked glycoprotein in the p62 complex from rat tissues. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction