Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP62 Monoclonal Antibody (RL1)
GREENER_CHOICE

Catalog No. MA1071
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1071 Supplier Invitrogen™ Supplier No. MA1071
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-071 detects nuclear pore-O-linked glycoprotein from rat, amphibian and yeast. MA1-071 has been successfully used in Western blot, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody detects up to eight different proteins from the nuclear pore complex (NPC) of approximately 210, 180, 145, 100, 63, 58, 54 and 45 kDa. Immunofluorescence staining of NPC O-linked glycoproteins with MA1-071 results in exclusive labeling of the NPC proteins on a wide variety of mammalian cells as well as S. cerevisiae and Xenopus. Labeling occurs exclusively at the NPC with most of the labeling at the cytoplasmic and nucleoplasmic margins. Microinjected MA1-071 inhibits both protein import and RNA export in Xenopus oocytes. The MA1-071 immunogen is pore complex-lamina fraction purified from rat liver nuclear envelopes.

SDS3 is a subunit of the histone deacetylase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP62
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RL1
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Nup62
Gene Accession No. P17955
Gene Alias 62 kDa nucleoporin; AA589433; AI426861; AI790512; AU045898; D7Ertd649e; DKFZp547L134; FLJ20822; FLJ43869; IBSN; MGC841; Np62; nuclear pore complex 1; nuclear pore glycoprotein 62; nuclear pore glycoprotein p62; nucleoporin 62; nucleoporin 62kD; nucleoporin 62kDa; nucleoporin Nup62; nucleoporin p62; Nup62; Nupc1; p62; SNDI
Gene Symbols Nup62
Host Species Mouse
Immunogen Pore complex-lamina fraction purified from rat liver nuclear envelopes.
Purification Method PEG precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65274
Target Species Rat, Amphibian, Yeast
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.