Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP62 Mouse anti-Human, Mouse, Rat, Clone: 2G12B3, Proteintech
SDP

Catalog No. 501736991 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6991 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6991 Supplier Proteintech Group Inc Supplier No. 665731IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SDS3 is a subunit of the histone deacetylase.

Specifications

Antigen NUP62
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2G12B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NUP62
Gene Accession No. P17955, P37198, Q63850
Gene Alias 62 kDa nucleoporin, IBSN, Nuclear pore glycoprotein p62, nucleoporin 62kDa, Nucleoporin Nup62, NUP62, p62, SNDI
Gene Symbols Nup62
Host Species Mouse
Immunogen NUP62 Fusion Protein Ag4912
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18226, 23636, 65274
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.