Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP62 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142089
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142089 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142089 Supplier Invitrogen™ Supplier No. PA5142089
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

SDS3 is a subunit of the histone deacetylase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP62
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Nup62
Gene Accession No. P37198
Gene Alias 62 kDa nucleoporin; AA589433; AI426861; AI790512; AU045898; D7Ertd649e; DKFZp547L134; FLJ20822; FLJ43869; IBSN; MGC841; Np62; nuclear pore complex 1; nuclear pore glycoprotein 62; nuclear pore glycoprotein p62; nucleoporin 62; nucleoporin 62kD; nucleoporin 62kDa; nucleoporin Nup62; nucleoporin p62; Nup62; Nupc1; p62; SNDI
Gene Symbols Nup62
Host Species Goat
Immunogen Peptide with sequence C-RKEQERSFRITFD.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23636
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.