Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP62 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582187
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582187 Supplier Invitrogen™ Supplier No. PA582187
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGFTFGTAKT ATTTPATGFS FSTSGTGGFN FGAPFQPATS TPSTGLFSLA TQTPATQTTG FTFGTATLAS GGTGFSLGIG ASKLNLSNTA ATPAMANPSG FGLGSSNLTN AISSTVTSSQ GTAPTGFVFG PST Highest antigen sequence indentity to the following orthologs: Rat - 79%, Mouse - 79%.

SDS3 is a subunit of the histone deacetylase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP62
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nup62
Gene Accession No. P37198
Gene Alias 62 kDa nucleoporin; AA589433; AI426861; AI790512; AU045898; D7Ertd649e; DKFZp547L134; FLJ20822; FLJ43869; IBSN; MGC841; Np62; nuclear pore complex 1; nuclear pore glycoprotein 62; nuclear pore glycoprotein p62; nucleoporin 62; nucleoporin 62kD; nucleoporin 62kDa; nucleoporin Nup62; nucleoporin p62; Nup62; Nupc1; p62; SNDI
Gene Symbols Nup62
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUP62. Recombinant protein control fragment (Product #RP-89199).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23636
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.