Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP62 Rabbit anti-Human, Mouse, Rat, Clone: 4V10S5, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315729100UL
Description
NUP62 Monoclonal antibody specifically detects NUP62 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| NUP62 | |
| Monoclonal | |
| Unconjugated | |
| DKFZp547L134,62 kDa nucleoporin, FLJ20822, FLJ43869, IBSNp62, MGC841, nuclear pore glycoprotein p62, nucleoporin 62kD, nucleoporin 62kDa, SNDINucleoporin Nup62 | |
| A synthetic peptide corresponding to a sequence within amino acids 423-522 of human NUP62 (P37198). | |
| 100 μg | |
| Apoptosis, Cell Biology, Signal Transduction | |
| 23636 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 4V10S5 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction