Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP62 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | NUP62 |
|---|---|
| Concentration | 0.27 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9088
![]()
|
Proteintech Group Inc
139161AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
SDS3 is a subunit of the histone deacetylase.Specifications
| NUP62 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P17955, P37198, Q63850 | |
| 18226, 23636, 65274 | |
| NUP62 Fusion Protein Ag4912 | |
| Primary | |
| -20°C | |
| NUP62 |
| 0.27 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 62 kDa nucleoporin, IBSN, Nuclear pore glycoprotein p62, nucleoporin 62kDa, Nucleoporin Nup62, NUP62, p62, SNDI | |
| Nup62 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title