Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP85 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584557 Supplier Invitrogen™ Supplier No. PA584557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGEPTVTLIP GVNSKKNQMY FDWGPGEMLV CETSFNKKEK SEMVPSCPFI YIIRKDVDVY SQILRKLFNE SHGIFL Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 87%.

Bidirectional transport of macromolecules between the cytoplasm and nucleus occurs through nuclear pore complexes (NPCs) embedded in the nuclear envelope. NPCs are composed of subcomplexes, and NUP85 is part of one such subcomplex, Nup107-160.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP85
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nup85
Gene Accession No. Q9BW27
Gene Alias 85 kDa nucleoporin; FLJ12549; FROUNT; Nuclear pore complex protein Nup85; nucleoporin 85; nucleoporin 85kDa; nucleoporin Nup75; Nucleoporin Nup85; NUP75; Nup85; PCN; PCNT; Pcnt1; pericentrin 1; pericentrin-1
Gene Symbols Nup85
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUP85. Recombinant protein control fragment (Product #RP-101987).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79902
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.