Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUP85 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584522
Description
Immunogen sequence: ENPSKHDSFW NLVTILVLQG RLDEARQMLS KEADASPASA GICRIMGDLM RTMPILSPGN TQTLTELELK WQ.
Bidirectional transport of macromolecules between the cytoplasm and nucleus occurs through nuclear pore complexes (NPCs) embedded in the nuclear envelope. NPCs are composed of subcomplexes, and NUP85 is part of one such subcomplex, Nup107-160.
Specifications
NUP85 | |
Polyclonal | |
Unconjugated | |
Nup85 | |
85 kDa nucleoporin; FLJ12549; FROUNT; Nuclear pore complex protein Nup85; nucleoporin 85; nucleoporin 85kDa; nucleoporin Nup75; Nucleoporin Nup85; NUP75; Nup85; PCN; PCNT; Pcnt1; pericentrin 1; pericentrin-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79902 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BW27 | |
Nup85 | |
Recombinant protein corresponding to Human NUP85. Recombinant protein control fragment (Product #RP-105215). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction