Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP85 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588186 Supplier Invitrogen™ Supplier No. PA588186
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CPELGRVSLE LHIERIPLNT EQKALKVLRI CEQRQMTEQV RSICKILAMK AVRNNRLGSA LSWSIRAKDA AFATLVSDRF LRDYCERGCF SDLDLIDNLG PAMMLSDRLT FLGKYREFHR MYGEKRFADA ASLLLSLMTS RIAPRSFWMT LLTDALPLLE QKQVIFSAEQ TYELMRCLED LTSRRPVHGE SDTEQLQDDD IETTKVEMLR LSLARNLARA IIREGSLEGS; Positive Samples: HT-29, U-87MG, A-549, LO2, HeLa, Mouse brain, Mouse testis, Rat testis; Cellular Location: Chromosome, Cytoplasm, Nucleus, Nucleus membrane, centromere, cytoskeleton, kinetochore, nuclear pore complex, spindle.

Bidirectional transport of macromolecules between the cytoplasm and nucleus occurs through nuclear pore complexes (NPCs) embedded in the nuclear envelope. NPCs are composed of subcomplexes, and NUP85 is part of one such subcomplex, Nup107-160.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP85
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Nup85
Gene Accession No. Q4QQS8, Q8R480, Q9BW27
Gene Alias 85 kDa nucleoporin; FLJ12549; FROUNT; Nuclear pore complex protein Nup85; nucleoporin 85; nucleoporin 85kDa; nucleoporin Nup75; Nucleoporin Nup85; NUP75; Nup85; PCN; PCNT; Pcnt1; pericentrin 1; pericentrin-1
Gene Symbols Nup85
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 427-656 of human NUP85 (NP_0791201).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287830, 445007, 79902
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.