Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUP85 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598920
Description
NUP85 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Bidirectional transport of macromolecules between the cytoplasm and nucleus occurs through nuclear pore complexes (NPCs) embedded in the nuclear envelope. NPCs are composed of subcomplexes, and NUP85 is part of one such subcomplex, Nup107-160.
Specifications
NUP85 | |
Polyclonal | |
Unconjugated | |
Nup85 | |
85 kDa nucleoporin; FLJ12549; FROUNT; Nuclear pore complex protein Nup85; nucleoporin 85; nucleoporin 85kDa; nucleoporin Nup75; Nucleoporin Nup85; NUP75; Nup85; PCN; PCNT; Pcnt1; pericentrin 1; pericentrin-1 | |
Rabbit | |
Protein G | |
RUO | |
445007, 79902 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8R480, Q9BW27 | |
Nup85 | |
Recombinant Human Nuclear pore complex protein Nup85 protein (201-500AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction