Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUP88 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554518
Description
Immunogen sequence: QLLTRNVVFG LGGELFLWDG EDSSFLVVRL RGPSGGGEEP ALSQYQRLLC INPPLFEIYQ VLLSPTQHHV ALIGIKGLMV LELPKRWGKN SEFEGGKSTV NCSTTPVAER FFTSSTSLTL KHAAWYPSEI LDPHVV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.
The nuclear pore complex (NPC) mediates bidirectional macromolecular traffic between the nucleus and cytoplasm in eukaryotic cells and is comprised of more than 100 different subunits. Many of the subunits belong to a family called nucleoporins (Nups), which are characterized by the presence of O-linked-N-acetylglucosamine moieties and a distinctive pentapeptide repeat (XFXFG). Nup88 (nucleoporin 88 kDa) is a 741 amino acid protein that localizes to the nucleus and functions as an essential component of the nuclear pore complex. Expressed ubiquitously, Nup88 is subject to phosphorylation by ATM or ATR and is upregulated in malignant neoplasms and precancerous dysplasias, suggesting a role in tumorigenesis. The gene encoding Nup88 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1, 200 genes.
Specifications
| NUP88 | |
| Polyclonal | |
| Unconjugated | |
| NUP88 | |
| 88 kDa nuclear pore complex protein; 88 kDa nucleoporin; karyoporin; nuclear pore complex protein 88; nuclear pore complex protein Nup88; nucleoporin 88; nucleoporin 88kDa; nucleoporin Nup84; Nucleoporin Nup88; Nup84; NUP88; Prei2; preimplantation protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4927 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99567 | |
| NUP88 | |
| Recombinant protein corresponding to Human NUP88. Recombinant protein control fragment (Product #RP-93198). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction