Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP93 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP181546
Description
NUP93 Polyclonal specifically detects NUP93 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NUP93 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Q8N1F7 | |
NUP93 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VERNLQEIQQAGERLRSRTLTRTSQETADVKASVLLGSRGLDISHISQRLESLSAATTFEPLEPVKDTDIQGFLKNEKDNALLSAIEESRKRTFGMAEEYHRESMLVEWEQVKQRI | |
Affinity Purified | |
RUO | |
9688 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
KIAA0095Nucleoporin Nup93,93 kDa nucleoporin, MGC21106, nuclear pore complex protein Nup93, nucleoporin 93kDa | |
Rabbit | |
93 kDa | |
0.1 mL | |
Primary | |
Specificity of human NUP93 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction