Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUP98 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093327C
Description
NUP98 Polyclonal antibody specifically detects NUP98 in Human samples. It is validated for Western BlotSpecifications
NUP98 | |
Polyclonal | |
Western Blot | |
ADAR2, ADIR2, GLFG-repeat containing nucleoporin, nuclear pore complex protein Nup98-Nup96, nucleoporin 98kD, nucleoporin 98kDa, NUP196, NUP96, Nup98-Nup96 | |
The immunogen recognized by this antibody maps to a region between residue 600 and 650 of human nucleoporin 98kDa using the numbering given in entry NP_057404.2 (GeneID 4928). | |
0.1 mL | |
Cancer, Mitotic Regulators | |
4928 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction