Learn More
NUP98-NUP96 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 123291AP150UL

Description
This antibody was raised against the carboxy-terminal region of Nup98-Nup96, it recognizes all Nup98-Nup96 proteins except Nup98-N. Since the autoproteolysis is very rapid this antibody usually detected 98-115 kDa Nup96 protein in multiple cell lines. It
ZFP64 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers. ZFP64 may be involved in transcriptional regulation.Specifications
NUP98-NUP96 | |
Polyclonal | |
Unconjugated | |
NUP98 | |
96 kDa nucleoporin, ADAR2, ADIR2, Nucleoporin Nup96, NUP196, NUP96, NUP98 NUP96, NUP98-NUP96 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
269966, 4928, 81738 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49793, P52948, Q6PFD9 | |
NUP98 | |
NUP98-NUP96 Fusion Protein Ag3016 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.