Learn More
Invitrogen™ NUP98 Polyclonal Antibody

Description
It is not recommended to aliquot this antibody.
Specifications
Specifications
| Antigen | NUP98 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 |
| Gene | NUP98 |
| Gene Accession No. | P49793, P52948, Q6PFD9 |
| Gene Alias | 4732457F17; 96 kDa nucleoporin; 98 kDa nucleoporin; ADAR2; adir2; AI849286; GLFG-repeat containing nucleoporin; im:7151238; LOW QUALITY PROTEIN: nuclear pore complex protein Nup98-Nup96; Nuclear pore complex protein Nup96; Nuclear pore complex protein Nup98; nuclear pore complex protein Nup98; nuclear pore complex protein Nup98-Nup96; nuclear pore complex protein Nup98-Nup96; nucleoporin 98; nucleoporin 98 and 96 precursor; nucleoporin 98 L homeolog; nucleoporin 98kD; nucleoporin 98kDa; Nucleoporin Nup96; Nucleoporin Nup98; NUP196; Nup96; NUP98; nup98.L; NUP98/PHF23 fusion 2 protein; Nup98-Nup96; OTTHUMP00000013954; zgc:113968; zgc:63593; zgc:63593 protein |
| Gene Symbols | NUP98, nup98.L |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.