Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP98 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595227
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595227 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595227 Supplier Invitrogen™ Supplier No. PA595227
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Human NUP98 shares 95% and 94% amino acid (aa) sequence identity with mouse and rat NUP98, respectively. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human K562 whole cell, human MCF-7 whole cell, human COLO320 whole cell, human HepG2 whole cell, human Hacat whole cell, monkey COS-7 whole cell, rat NRK whole cell, mouse lung tissue, mouse NIH/3T3 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Signal-mediated nuclear import and export proceed through the nuclear pore complex (NPC), which is comprised of approximately 50 unique proteins collectively known as nucleoporins. The 98 kDa nucleoporin is generated through a biogenesis pathway that involves synthesis and proteolytic cleavage of a 186 kDa precursor protein. This cleavage results in the 98 kDa nucleoporin as well as a 96 kDa nucleoporin, both of which are localized to the nucleoplasmic side of the NPC. Rat studies show that the 98 kDa nucleoporin functions as one of several docking site nucleoporins of transport substrates. The human gene has been shown to fuse to several genes following chromosome translocations in acute myelogenous leukemia (AML) and T-cell acute lymphocytic leukemia (T-ALL). This gene is one of several genes located in the imprinted gene domain of 11p15. 5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in several transcript variants; however, not all variants have been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP98
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.01mg sodium azide
Gene NUP98
Gene Accession No. P49793, P52948, Q6PFD9
Gene Alias 4732457F17; 96 kDa nucleoporin; 98 kDa nucleoporin; ADAR2; adir2; AI849286; GLFG-repeat containing nucleoporin; im:7151238; LOW QUALITY PROTEIN: nuclear pore complex protein Nup98-Nup96; Nuclear pore complex protein Nup96; Nuclear pore complex protein Nup98; nuclear pore complex protein Nup98; nuclear pore complex protein Nup98-Nup96; nuclear pore complex protein Nup98-Nup96; nucleoporin 98; nucleoporin 98 and 96 precursor; nucleoporin 98 L homeolog; nucleoporin 98kD; nucleoporin 98kDa; Nucleoporin Nup96; Nucleoporin Nup98; NUP196; Nup96; NUP98; nup98.L; NUP98/PHF23 fusion 2 protein; Nup98-Nup96; OTTHUMP00000013954; zgc:113968; zgc:63593; zgc:63593 protein
Gene Symbols NUP98
Host Species Rabbit
Immunogen E.coli-derived human NUP98 recombinant protein (Position: H549-F880).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269966, 4928, 81738
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.