Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP98 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. PIPA5121732
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121732 100 μL
PIPA5121733 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121732 Supplier Invitrogen™ Supplier No. PA5121732
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Signal-mediated nuclear import and export proceed through the nuclear pore complex (NPC), which is comprised of approximately 50 unique proteins collectively known as nucleoporins. The 98 kDa nucleoporin is generated through a biogenesis pathway that involves synthesis and proteolytic cleavage of a 186 kDa precursor protein. This cleavage results in the 98 kDa nucleoporin as well as a 96 kDa nucleoporin, both of which are localized to the nucleoplasmic side of the NPC. Rat studies show that the 98 kDa nucleoporin functions as one of several docking site nucleoporins of transport substrates. The human gene has been shown to fuse to several genes following chromosome translocations in acute myelogenous leukemia (AML) and T-cell acute lymphocytic leukemia (T-ALL). This gene is one of several genes located in the imprinted gene domain of 11p15. 5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in several transcript variants; however, not all variants have been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP98
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene NUP98
Gene Accession No. P52948, Q6PFD9
Gene Alias 4732457F17; 96 kDa nucleoporin; 98 kDa nucleoporin; ADAR2; adir2; AI849286; GLFG-repeat containing nucleoporin; im:7151238; LOW QUALITY PROTEIN: nuclear pore complex protein Nup98-Nup96; Nuclear pore complex protein Nup96; Nuclear pore complex protein Nup98; nuclear pore complex protein Nup98; nuclear pore complex protein Nup98-Nup96; nuclear pore complex protein Nup98-Nup96; nucleoporin 98; nucleoporin 98 and 96 precursor; nucleoporin 98 L homeolog; nucleoporin 98kD; nucleoporin 98kDa; Nucleoporin Nup96; Nucleoporin Nup98; NUP196; Nup96; NUP98; nup98.L; NUP98/PHF23 fusion 2 protein; Nup98-Nup96; OTTHUMP00000013954; zgc:113968; zgc:63593; zgc:63593 protein
Gene Symbols NUP98
Host Species Rabbit
Immunogen Gln629-Pro894 (Accession P52948), with N-terminal His Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269966, 4928
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.