Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NuPAGE™ Bis-Tris Welcome Pack, 12%

Catalog No. p-7224321
Encompass_Preferred
Click to view available options
Wells:
10-well
12-well
15-well

The NuPAGE Bis-Tris Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the Mini Gel Tank.

The NuPAGE Bis-Tris Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the Mini Gel Tank. The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies used.

The NuPAGE Bis-Tris Welcome Pack contains:
• Mini Gel Tank (A25977)
• NuPAGE Bis-Tris mini gels (2 boxes, 20 gels)
• NuPAGE MES SDS Running Buffer, 20X (NP0002)
• NuPAGE LDS Sample Buffer, 4X (NP0007)
• NuPAGE Sample Reducing Agent, 10X (NP0009)
• PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)

About the Mini Gel Tank
The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies used.

About NuPAGE Bis-Tris gels
Invitrogen NuPAGE Bis-Tris Protein gels are precast polyacrylamide gels designed to give optimal separation of a wide range of proteins under denaturing conditions. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is crucial. Also, use NuPAGE gels for optimal results during day-to-day use.

Transfer
For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer Buffer (NP0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Warranty and Services

Up to 12 months

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mini Gel Tank
Gel Percentage 12%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
Separation Range 3.5 to 80 kDa
Wells 10-well
Product Line NuPAGE™
Quantity 1 Welcome Pk. kit
Shipping Condition Approved for shipment at room temperature and wet ice
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.