Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NuPAGE™ Tris-Acetate Welcome Pack, 7%

Catalog No. p-7224314
Encompass_Preferred
Click to view available options
Wells:
10-well
12-well
15-well

The NuPAGE Tris-Acetate Welcome Pack provides all of the necessary NuPAGE Tris-Acetate gels, buffers, and reagents needed to begin using the Mini Gel Tank.

The NuPAGE Tris-Acetate Welcome Pack provides all of the necessary NuPAGE Tris-Acetate gels, buffers, and reagents you will need to begin using the Mini Gel Tank. The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading, and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies used.

The NuPAGE Tris-Acetate Welcome Pack contains:
• Mini Gel Tank
• NuPAGE Tris-Acetate Mini gels (2 boxes, 20 gels)
• NuPAGE Tris-Acetate SDS Running Buffer (20X)
• NuPAGE LDS Sample Buffer (4X)
• NuPAGE Sample Reducing Agent (10X)
• HiMark™ Pre-stained Protein Standard

Mini Gel Tank
The Mini Gel Tank is compatible with all Novex, NuPAGE and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies being used.

NuPAGE Tris-Acetate Gels
NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. NuPAGE Tris-Acetate protein gels are high performance polyacrylamide gels that simulate the denaturing or the native conditions of the traditional laemmli system. Its unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins compared to traditional Tris-glycine SDS-PAGE gels.

Transfer and detection
NuPAGE Tris-Acetate gels are compatible with the Mini Blot Module, Pierce Power Blotter, iBlot 2 Gel Transfer Device, iBind Western Device, and iBind Flex Western Device for optimal transfer and detection of proteins.

Warranty and Services

Up to 12 months

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mini Gel Tank
Gel Percentage 7%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tris-Acetate
Separation Range 36 to 500 kDa
Wells 10-well
Product Line NuPAGE™
Quantity 1 Welcome Pk. kit
Shipping Condition Approved for shipment at room temperature and wet ice
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.