Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUPL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558753
Description
Immunogen sequence: RNTLNIDKLK IETALELKNA EIALRTQKTP PGLQHEYAAP ADYFRILVQQ FEVQLQQYRQ QIEELENHLA TQANNSHITP QDLSMAMQKI Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
This gene encodes a member of the nucleoporin family that shares 87% sequence identity with rat nucleoporin p58. The protein is localized to the nuclear rim and is a component of the nuclear pore complex (NPC). All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
NUPL1 | |
Polyclonal | |
Unconjugated | |
NUP58 | |
1700017F11Rik; 58 kDa nucleoporin; KIAA0410; mKIAA0410; nucleoporin 58; nucleoporin 58kDa; nucleoporin like 1; nucleoporin p45; nucleoporin p58; nucleoporin p58/p45; nucleoporin-like 1; nucleoporin-like protein 1; NUP45; NUP58; Nupl1; p58/p45; PRO2463 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9818 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BVL2 | |
NUP58 | |
Recombinant protein corresponding to Human NUPL1. Recombinant protein control fragment (Product #RP-97971). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction