Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUPL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110053
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110053 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110053 Supplier Invitrogen™ Supplier No. PA5110053
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAQLQSIHEN VKVLKEQYLG YRKMFLGDAV DVFETRRAEA KKWQNTPRVT TGPTPFSTMP NAAAVAMAAT LTQQQQPATG PQPSLGVSFG TPFGSGIGTG L.

This gene encodes a member of the nucleoporin family that shares 87% sequence identity with rat nucleoporin p58. The protein is localized to the nuclear rim and is a component of the nuclear pore complex (NPC). All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUPL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NUP58
Gene Accession No. P70581
Gene Alias 1700017F11Rik; 58 kDa nucleoporin; KIAA0410; mKIAA0410; nucleoporin 58; nucleoporin 58kDa; nucleoporin like 1; nucleoporin p45; nucleoporin p58; nucleoporin p58/p45; nucleoporin-like 1; nucleoporin-like protein 1; NUP45; NUP58; Nupl1; p58/p45; PRO2463
Gene Symbols NUP58
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 400-500 of human NUP58 (NP_0548081).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245922
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.