Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUPL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113478
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113478 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113478 Supplier Invitrogen™ Supplier No. PA5113478
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUPL1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a member of the nucleoporin family that shares 87% sequence identity with rat nucleoporin p58. The protein is localized to the nuclear rim and is a component of the nuclear pore complex (NPC). All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUPL1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NUP58
Gene Accession No. Q8R332, Q9BVL2
Gene Alias 1700017F11Rik; 58 kDa nucleoporin; KIAA0410; mKIAA0410; nucleoporin 58; nucleoporin 58kDa; nucleoporin like 1; nucleoporin p45; nucleoporin p58; nucleoporin p58/p45; nucleoporin-like 1; nucleoporin-like protein 1; NUP45; NUP58; Nupl1; p58/p45; PRO2463
Gene Symbols NUP58, Nupl1
Host Species Rabbit
Immunogen Recombinant Human Nucleoporin p58/p45 protein (211-480AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71844, 9818
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.