Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUPL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984342
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4342 Supplier Bioss Supplier No. BS19569R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen NUPL1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NUP58
Gene Accession No. Q9BVL2
Gene Alias 1700017F11Rik; 58 kDa nucleoporin; KIAA0410; mKIAA0410; nucleoporin 58; nucleoporin 58kDa; nucleoporin like 1; nucleoporin p45; nucleoporin p58; Nucleoporin p58/p45; nucleoporin-like 1; Nucleoporin-like protein 1; NUP45; NUP58; NUPL1; p58/p45; PRO2463
Gene Symbols NUP58
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NUPL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9818
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.