Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588035
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588035 Supplier Invitrogen™ Supplier No. PA588035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAICQFFLQG RCRFGDRCWN EHPGARGAGG GRQQPQQQPS GNNRRGWNTT SQRYSNVIQP SSFSKSTPWG GSRDQEKPYF SSFDSGASTN RKEGFGLSEN PFASLSPDEQ KDEKKLLEGI VKDMEVWESS GQWMFSVYSP VKKKPNISGF TDISPEELRL EYHNFLTSNN LQSYLNSVQR LINQWRNRVN ELKSLNISTK; Positive Samples: HeLa, A-549, A-431, SGC-7901; Cellular Location: Cytoplasmic side, Nucleus, Nucleus membrane, Peripheral membrane protein, nuclear pore complex.

Required for the export of mRNAs containing poly(A) tails from the nucleus into the cytoplasm. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUPL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NUP42
Gene Accession No. O15504
Gene Alias AV116043; CG1; h H_RG271G13.9; H_RG271G13.9; hCG1; NLP_1; Nlp1; NLP-1; nucleoporin hCG1; nucleoporin like 2; Nucleoporin NUP42; Nucleoporin-42; nucleoporin-like protein 1; nucleoporin-like protein 2; NUP42; NUP42 homolog; Nupl2; RGD1560934
Gene Symbols NUP42
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human NUPL2 (NP_0313681).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11097
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.