Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUPR1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501092790 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-109-2790 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-109-2790 Supplier Proteintech Group Inc Supplier No. 150561AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The human p8 gene encodes a nuclear protein with strong mitogenic activity, which is expressed in the pancreas in response to acute pancreatitis. Response to TNF alpha and staurosporine-induced apoptosis is not modified by p8 expression. However, cells expressing p8 grow significantly and more rapidly.

Specifications

Antigen NUPR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NUPR1
Gene Accession No. O60356
Gene Alias Candidate of metastasis 1, COM1, nuclear protein 1, NUPR1, P8, Protein p8
Gene Symbols NUPR1
Host Species Rabbit
Immunogen NUPR1 Fusion Protein Ag7039
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26471
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.