Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nur77 Mouse anti-Mouse, Human, Alexa Fluor™ 647, Clone: 12.14, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566735
Description
Nur77 Monoclonal antibody specifically detects Nur77 in Mouse,Human samples. It is validated for Flow Cytometry.
Specifications
Nur77 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Nur77 | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
12.14 | |
Alexa Fluor 647 | |
0.1 mg | |
Primary | |
Mouse, Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction