Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nurim Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675520
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-520 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-520 Supplier Invitrogen™ Supplier No. PA141391
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: HeLa, NIH-3T3, PC3. Predicted to react with Chimpanzee and Rhesus Monkey.

Nurim is an inner nuclear membrane (INM) protein that is tightly associated with the inner nuclear membrane. It was first isolated in a visual screen for nuclear envelope-localizing proteins. In subcellular fractionation, nurim remained extremely tightly bound to nuclear fractions. Unlike the known nuclear envelope (NE) membrane proteins, it is neither associated with nuclear pores, nor targeted like lamin-associated membrane proteins suggesting its localization to NE by a distinct mechanism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nurim
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene NRM
Gene Accession No. Q8IXM6, Q8VC65
Gene Alias 2610307M02Rik; AI429796; DAAP-285E11.8; Nrm; NRM29; Nuclear envelope membrane protein; nuclear rim protein; Nurim; nurim (nuclear envelope membrane protein); UNQ555/PRO1112
Gene Symbols NRM
Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human Nurim.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106582, 11270
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.