Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nurr-1 Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN167525UL
Description
Anti-Nurr-1 is a highly specific rabbit polyclonal antibody that targets Nuclear receptor subfamily 4 group A member 2 and has been tested in ELISA, Immunohistochemistry (Paraffin), and Western Blotting.
Specifications
Nurr-1 | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
Mouse, Human | |
Semi-purified |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
Please refer to lot specific datasheet. | |
NR4A2,NOT,NURR1,TINUR | |
His-tagged recombinant fragment corresponding to 271 amino acids from the C-terminal half of human Nurr-1 and includes the ligand binding domain. | |
25 μL | |
Neuroscience | |
This rabbit polyclonal antibody detects Nurr-1 in human and murine tissues. It targets an epitope within the C-terminal half. | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction