Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nurr1 Polyclonal Antibody, Bioss
SDP

Catalog No. 502279436
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-9436 Supplier Bioss Supplier No. BS0178R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Nurr1 Polyclonal antibody specifically detects Nurr1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot

Specifications

Antigen Nurr1
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene NR4A2
Gene Accession No. P43354, Q06219, Q07917
Gene Alias HZF3; Hzf-3; Immediate-early response protein NOT; intermediate-early receptor protein; NGFI-B; NGFI-B beta; NGFI-B/nur77 beta-type transcription factor homolog; NOT; NR4A2; Nuclear orphan receptor HZF-3; nuclear receptor related 1; nuclear receptor subfamily 4 group A member 2; nuclear receptor subfamily 4 group A member 2 variant TINUR; nuclear receptor subfamily 4 group A member 2 variant NURR1a; nuclear receptor subfamily 4 group A member 2 variant NURR1b; nuclear receptor subfamily 4 group A member 2 variant NURR1c; nuclear receptor subfamily 4 group A member 2 variant NURR2c; nuclear receptor subfamily 4, group A, member 2; nur related protein 1; nur related protein-1, human homolog of; Nurr1; NUR-related factor 1; orphan nuclear receptor NR4A2; orphan nuclear receptor NURR1; Regenerating liver nuclear receptor 1; RNR1; RNR-1; SL-322; T-cell nuclear receptor NOT; TINOR; TINUR; transcriptionally inducible nuclear receptor related 1; transcriptionally-inducible nuclear receptor
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Nurr1
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18227, 4929, 54278
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.