Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100653
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100653 Supplier Invitrogen™ Supplier No. PA5100653
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NUS1.

With DHDDS, forms the dehydrodolichyl diphosphate synthase (DDS) complex, an essential component of the dolichol monophosphate (Dol-P) biosynthetic machinery. Adds multiple copies of isopentenyl pyrophosphate (IPP) to farnesyl pyrophosphate (FPP) to produce dehydrodolichyl diphosphate (Dedol-PP), a precursor of dolichol which is utilized as a sugar carrier in protein glycosylation in the endoplasmic reticulum (ER). Regulates the glycosylation and stability of nascent NPC2, thereby promoting trafficking of LDL-derived cholesterol. Acts as a specific receptor for the N-terminus of Nogo-B, a neural and cardiovascular regulator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUS1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NUS1
Gene Accession No. Q96E22, Q99LJ8
Gene Alias 1600027K07Rik; AU019165; AW538011; BC003223; C6orf68; Cis-prenyltransferase subunit NgBR; D10Ertd438e; dehydrodolichyl diphosphate syntase complex subunit NUS1; dehydrodolichyl diphosphate syntase complex subunit NUS1 pseudogene; dehydrodolichyl diphosphate synthase complex subunit NUS1; di-trans,poly-cis-decaprenylcistransferase; MGC:7199; NgBR; Nogo-B receptor; Nuclear undecaprenyl pyrophosphate synthase 1 homolog; nuclear undecaprenyl pyrophosphate synthase 1 homolog (S. cerevisiae); Nus1; NUS1 dehydrodolichyl diphosphate synthase subunit; NUS1, dehydrodolichyl diphosphate synthase subunit; RGD1307879; TANGO14; transport and golgi organization 14 homolog
Gene Symbols NUS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NUS1(Accession Q96E22), corresponding to amino acid residues N145-D195.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116150, 294400, 52014
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.