Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUTF2 Monoclonal Antibody (J.206.1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA514809
Description
It is not recommended to aliquot this antibody.
The protein encoded by this gene is a cytosolic factor that facilitates protein transport into the nucleus. It interacts with the nuclear pore complex glycoprotein p62. This encoded protein acts at a relative late stage of nuclear protein import, subsequent to the initial docking of nuclear import ligand at the nuclear envelope. It is thought to be part of a multicomponent system of cytosolic factors that assemble at the pore complex during nuclear import.
Specifications
NUTF2 | |
Monoclonal | |
400 μg/mL | |
0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
P61970, P61971, P61972 | |
NUTF2 | |
Full-length recombinant human NTF2. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG2a |
Western Blot, Immunocytochemistry | |
J.206.1 | |
Unconjugated | |
NUTF2 | |
2700067I02Rik; AI115459; AI413596; AW546000; Ntf2; NTF-2; Ntf3; nuclear transport factor 2; NUTF2; Placental protein 15; PP15 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10204, 291981, 68051 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction