Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUTF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559350
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559350 Supplier Invitrogen™ Supplier No. PA559350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KIQHSITAQD HQPTPDSCII SMVVGQLKAD EDPIMGFHQM FLLKNINDAW VCTNDMFRLA LHNFG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The protein encoded by this gene is a cytosolic factor that facilitates protein transport into the nucleus. It interacts with the nuclear pore complex glycoprotein p62. This encoded protein acts at a relative late stage of nuclear protein import, subsequent to the initial docking of nuclear import ligand at the nuclear envelope. It is thought to be part of a multicomponent system of cytosolic factors that assemble at the pore complex during nuclear import.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUTF2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUTF2
Gene Accession No. P61970
Gene Alias 2700067I02Rik; AI115459; AI413596; AW546000; Ntf2; NTF-2; Ntf3; nuclear transport factor 2; NUTF2; Placental protein 15; PP15
Gene Symbols NUTF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUTF2. Recombinant protein control fragment (Product #RP-98618).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10204
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.