Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Nutlin-3
GREENER_CHOICESDP

Catalog No. 398410 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-841-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 39-841-0 Supplier R&D Systems™ Supplier No. 3984/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. MDM2 antagonist; inhibits MDM2-p53 interaction

MDM2 antagonist; inhibits the MDM2-p53 interaction (IC50 = 0.09 μM) and activates p53. Antiproliferative agent; induces apoptosis in cancer cells.

Chemical Identifiers

CAS 548472-68-0
Molecular Formula C30H30Cl2N4O4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 581.49
Purity >98%
Molecular Weight (g/mol) 581.494
Chemical Name or Material Nutlin-3
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.