Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Nutlin 3a
GREENER_CHOICESDP

Catalog No. 60755 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-755 5 mg
60-752-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-755 Supplier Tocris Bioscience™ Supplier No. 6075/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

MDM2 antagonist; active enantiomer of Nutlin-3 (Cat. No. 3984)

Nutlin 3a is an MDM2 antagonist (IC50 = 90 nM); inhibits the MDM2-p53 interaction. Active enantiomer of Nutlin-3 (Cat. No. 3984). Induces expression for P53 regulated genes. Suppresses tumor growth in vivo >98% in prostate cancer and osteosarcoma cancer models. Antiproliferative.

Specifications

Chemical Name or Material 4-[[(4S,5R)-4,5-Bis(4-chlorophenyl)-4,5-dihydro-2-[4-methoxy-2-(1-methylethoxy)phenyl]-1H-imidazol-1-yl]carbonyl]-2-piperazinone
CAS 675576-98-4
Assay Percent Range >97%
Quantity 5 mg
Molecular Formula C30H30Cl2N4O4
Formula Weight Observed MW: 581.49
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.