Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUTM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559171
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559171 Supplier Invitrogen™ Supplier No. PA559171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TCPLNVHSYD PQGEGRVDPD LSKPKNLAPL QESQESYTTG TPKATSSHQG LGSTLPRRGT RNAIVPRETS VSKTHRSA Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 53%.

Plays a role in the regulation of proliferation. Regulates TERT expression by modulating SP1 binding to TERT promoter binding sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUTM1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUTM1
Gene Accession No. Q86Y26
Gene Alias C15orf55; FAM22H; Nuclear protein in testis; NUT; NUT family member 1; NUT midline carcinoma family member 1; NUT midline carcinoma, family member 1; NUTM1
Gene Symbols NUTM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUTM1. Recombinant protein control fragment (Product #RP-98181).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256646
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.