Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Nutrient Agar
SCI_ED

Catalog No. p-7158857 Shop All Aldon Products
Click to view available options
Quantity:
125 mL
200 mL

High quality microbiological media at an economical price

  • Innovating Science™ Sterile Prepared Agar and Broth will bring quality back to the lab market
  • Case of 24
  • Specifications

    Certifications/Compliance Sterile
    Concentration 1
    Content And Storage Room Temperature
    Form Gel
    Format Bottle
    Manufacturing Origin United States
    Media Type Nutrient Agar
    Product Type Prepared Microbiology Media
    Sterility Sterile
    Target Organism Class Bacteria
    Test Type Microbial Detection
    For Use With (Application) Microbes
    For Use With (Equipment) Microbes, Educational
    Packaging Type Glass
    Quantity 125 mL
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.