Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nutrient Broth

Catalog No. r061582
Click to view available options
Quantity:
100/Pk.
20/Pk.
Format:
Tube
Manufacturing Origin:
United States
This item is not returnable. View return policy
This item is not returnable. View return policy

Reduce media preparation time and cultivate fewer fastidious microorganisms using this convenient, pre-filled, ready-to-use general purpose medium.

Reduce media preparation time and cultivate fewer fastidious microorganisms using convenient, pre-filled, ready-to-use Thermo Scientific™ Nutrient Broth.

Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes of Selenite Broth.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear liquid, light amber
Content And Storage 2°C to 8°C
Format Tube
Manufacturing Origin United States
Product Type Microbiology Media
Fill Volume 5 mL
Quantity 20/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.