Learn More
Invitrogen™ NVL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120878
Description
Positive test controls include: NCI-H460, MCF7, Mouse testis, Mouse liver. The target is usually found in the following locations: Nucleus, nucleolus, nucleoplasm. Immunogen sequence: CLGIPDEASR ERILQTLCRK LRLPQAFDFC HLAHLTPGFV GADLMALCRE AAMCAVNRVL MKLQEQQKKN PEMEDLPSKG VQEERLGTEP TSETQDELQR LLGLLRDQDP LSEEQMQGLC IELNDFIVAL SSVQPSAKRE GFVTVPNVTW ADIGALEDIR EELTMAILAP VRNPDQFKAL G.
Gasdermin D is a member of the gasdermin family. Members of this family appear to play a role in regulation of epithelial proliferation. Gasdermin D has been suggested to act as a tumor suppressor. Alternatively spliced transcript variants have been described.
Specifications
NVL | |
Polyclonal | |
Unconjugated | |
NVL | |
1200009I24Rik; Nuclear valosin-containing protein-like; nuclear VCP-like; nuclear VCP-like protein; Nvl; NVL2; NVLp | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4931, 67459 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.73 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15381, Q9DBY8 | |
NVL | |
Recombinant fusion protein containing a sequence corresponding to amino acids 340-520 of human NVL (NP_001230076.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.