Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NVL Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501568880
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8880 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8880 Supplier Bethyl Laboratories, Inc Supplier No. A304863A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Valosin containing protein (VCP), also designated TERA (for transitional endoplasmic reticulum ATPase), is a member of the AAA family of ATPases, which are involved in a variety of cellular activities. VCP is involved in a variety of membrane functions and in the regulation of the cell cycle. VCP associates with ubiquitinated IkB-a as well as with the 26S proteosome, indicating a potential role for VCP in the proteosome-mediated degradation of IkB-a. NVL (nuclear valosin-containing protein-like), also known as NVLp, is an 856 amino acid nuclear protein belonging to the AAA ATPase family. Implicated in ATP-dependent nuclear processes and ribosome synthesis, NVL exists as three alternatively spliced isoforms designated NVL isoform 1 (NVLp.2), NVL isoform 2 (NVLp.1) and NVL isoform 3. Widely expressed, NVL is found at highest levels in pancreas, retina, heart, skeletal muscle and placenta.

Specifications

Antigen NVL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene NVL
Gene Accession No. O15381
Gene Alias Nuclear valosin-containing protein-like, NVLp, Nuclear VCP-like protein
Gene Symbols NVL
Host Species Rabbit
Immunogen Between 75 and 125
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4931
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.