Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NVP 2
GREENER_CHOICESDP

Catalog No. 65355 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-355 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 65-355 Supplier Tocris Bioscience™ Supplier No. 6535/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective ATP-competitive CDK9 inhibitor

NVP 2 is a potent and selective ATP-competitive CDK9 inhibitor (IC50 = 0.5 nM). Exhibits selectivity for CDK9 over a panel of 468 kinases, but inhibits DYRK1B (IC50 = 350 nM), displays <90% binding to CDK7 (IC50 >10 μM) and CDK13. Inhibits proliferation of MOLT4 cells in vitro. Induces apoptosis.

Specifications

Chemical Name or Material 4-[[[5'-Chloro-2'-[[trans-4-[[(1R)-2-methoxy-1-methylethyl]amino]cyclohexyl]amino][2,4'-bipyridin]-6-yl]amino]methyl]tetrahydro-2H-pyran-4-carbonitrile
CAS 1263373-43-8
Quantity 5 mg
Molecular Formula C27H37ClN6O2
Formula Weight 513.07
Product Type Inhibitor
Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.