Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NVP ADW 742
GREENER_CHOICE

Catalog No. 524710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-471-0 10 mg
52-475-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-471-0 Supplier Tocris Bioscience™ Supplier No. 5247/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

ATP-competitive inhibitor of IGF1R

NVP ADW 742 is an ATP-competitive inhibitor of IGF1R (IC50 in the range of 0.1-0.2 μM). Also inhibits c-Kit kinase activity (IC50 in the range of 3-5 μM). Exhibits a synergistic effect with Imatinib (Cat. No. 5906), enhancing the sensitivity of multiple small cell lung cancer cell lines to the chemotherapeutics Etoposide (Cat. No. 1226) and Carboplatin (Cat. No. 2626).

Specifications

Quantity 10 mg
Product Type NVP ADW 742
Purity >99%
Molecular Weight (g/mol) 453.58
Inhibitors NVP ADW 742

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.