Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NVS MLLT-1
GREENER_CHOICESDP

Catalog No. 748250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-825-0 50 mg
74-821-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 74-825-0 Supplier Tocris Bioscience™ Supplier No. 7482/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective MLLT1/3 inhibitor

NVS-MLLT-1 is a selective inhibitor of YEATS domain of MLLT1 and MLLT3 (IC50 values are 150 and 254 nM for MLLT1 and MLLT3, respectively). It shows selectivity for YEATS2/4 domains and over all bromodomains (CERC2 IC50 > 40 μM).

Specifications

Chemical Name or Material N-[2-[[(2 S)-2-Methyl-1-pyrrolidinyl]methyl]-1 H-benzimidazol-6-yl]-6-isoquinolinecarboxamide
CAS 2561471-13-2
Quantity 50 mg
Target Bromodomain Inhibitors
Molecular Formula C23H23N5O
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.